1-Octadecylamine, 95%
1-Octadecylamine, 95%
1-Octadecylamine, 95%
Thermo Scientific Chemicals

1-Octadecylamine, 95%

CAS: 124-30-1 | C18H39N | 269.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L15458.14
also known as L15458-14
Price (USD)
117.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
117.00
Each
Chemical Identifiers
CAS124-30-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Assay (GC)≥94.0%
FormCrystals or powder or crystalline powder or flakes or prills or fused solid or clear liquid as melt
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Octadecylamine is used as asphalt emulsifier, lubricant and dispersants. It acts as a cationic surfactant and find applications is hair rinse bases, wood preservatives, textile softeners, dyeing auxiliaries and pigment grinding aids. It is used in boiler as an additive, which prevents the corrosion arises due to steam. It is a basic building block and used as an intermediate in the preparation of their amine derivatives, ethoxylates and amides. Metal oxide nano crystals having controlled size and shape are prepared by the thermal decomposition of metal nitrates with octadecylamine.

Solubility
Soluble in chloroform, alcohol, ether, benzene and acetone.Insoluble in water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

General References:

  1. Wang, S.; Zhang, c.; Li, Y.; Li, B.; Yang, Y. Chirality of Single-Handed Twisted Titania Tubular Nanoribbons Prepared Through Sol-gel Transcription. Chirality 2015, 27 (8), 543-550.
  2. Ng, Y. H.; Tay, S. W.; Hong, R. S.; Hong, L. In situ formation of reverse polymeric micelles in liquid alkanes to lodge alcohol micro-droplets. RSC Adv. 2015, 5 (27), 21033-21041.