1,4-Diphenyl-1-butanone, 98%
1,4-Diphenyl-1-butanone, 98%
1,4-Diphenyl-1-butanone, 98%
Thermo Scientific Chemicals

1,4-Diphenyl-1-butanone, 98%

CAS: 5407-91-0 | C16H16O | 224.303 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L15597.03
also known as L15597-03
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
33.10
Save 4.45 (13%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
33.10
Save 4.45 (13%)
Each
Chemical Identifiers
CAS5407-91-0
IUPAC Name1,4-diphenylbutan-1-one
Molecular FormulaC16H16O
InChI KeyGBUMEGLMTNAXOM-UHFFFAOYSA-N
SMILESO=C(CCCC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or granules
Assay (GC)≥97.5%
Melting Point (clear melt)51.5-57.5?C
Appearance (Color)White to pale yellow or pale pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only