2,3-Dihydro-1H-benz[de]isoquinoline, 97%
2,3-Dihydro-1H-benz[de]isoquinoline, 97%
2,3-Dihydro-1H-benz[de]isoquinoline, 97%
2,3-Dihydro-1H-benz[de]isoquinoline, 97%
Thermo Scientific Chemicals

2,3-Dihydro-1H-benz[de]isoquinoline, 97%

CAS: 22817-26-1 | C12H11N | 169.227 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L15613.MD
also known as L15613-MD
Price (USD)
60.65
Online Exclusive
66.80
Save 6.15 (9%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
66.80
Save 6.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS22817-26-1
IUPAC Name3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Molecular FormulaC12H11N
InChI KeyNBWCGDRKSOXZNI-UHFFFAOYSA-N
SMILESC1NCC2=C3C1=CC=CC3=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)96.0-105.0?C
Appearance (Color)Green to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only