2,2-Diphenylpropylamine hydrochloride, 98+%
2,2-Diphenylpropylamine hydrochloride, 98+%
2,2-Diphenylpropylamine hydrochloride, 98+%
Thermo Scientific Chemicals

2,2-Diphenylpropylamine hydrochloride, 98+%

CAS: 40691-66-5 | C15H18ClN | 247.766 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15615.06
also known as L15615-06
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS40691-66-5
IUPAC Namehydrogen 2,2-diphenylpropan-1-amine chloride
Molecular FormulaC15H18ClN
InChI KeyAASCJSPDUDWGGQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(CN)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only