Tri-n-octylphosphine, tech. 90%
Tri-n-octylphosphine, tech. 90%
Tri-n-octylphosphine, tech. 90%
Thermo Scientific Chemicals

Tri-n-octylphosphine, tech. 90%

CAS: 4731-53-7 | C24H51P | 370.646 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Catalog number L15744.AC
also known as L15744-AC
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.00
Save 8.35 (14%)
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.00
Save 8.35 (14%)
Each
Chemical Identifiers
CAS4731-53-7
IUPAC Nametrioctylphosphane
Molecular FormulaC24H51P
InChI KeyRMZAYIKUYWXQPB-UHFFFAOYSA-N
SMILESCCCCCCCCP(CCCCCCCC)CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥88.0%
CommentMay become cloudy on storage and form precipitate on standing
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Mntungwa, N.; Khan, M. D.; Mlowe, S.; Revaprasadu, N. A simple route to alkylamine capped antimony nanoparticles. Mater. Lett. 2015, 145, 239-242.
  2. More, D. S.; Moloto, M. J.; Moloto, N.; Matabola, K. P. TOPO-capped silver selenide nanoparticles and their incorporation into polymer nanofibers using electrospinning technique. Mater. Res. Bull. 2015, 65, 14-22.