12-Molybdophosphoric acid hydrate, Mo >47.5%
12-Molybdophosphoric acid hydrate, Mo >47.5%
12-Molybdophosphoric acid hydrate, Mo >47.5%
Thermo Scientific Chemicals

12-Molybdophosphoric acid hydrate, Mo >47.5%

CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L15827.36
also known as L15827-36
Price (USD)
392.65
Special offer
Online exclusive
Ends: 30-Jun-2026
462.00
Save 69.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
392.65
Special offer
Online exclusive
Ends: 30-Jun-2026
462.00
Save 69.35 (15%)
Each
Chemical Identifiers
CAS51429-74-4
IUPAC Name12-Molybdophosphoric acid
Molecular FormulaH3Mo12O40P
Molecular Weight (g/mol)1825.40
Synonymphosphoric acid dodecamolybdenum hexatriacontaoxidandiide
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green or yellow-orange
FormCrystals or powder or crystalline powder or adhering crystals
Elemental Analysis(Molybdenum, Mo) : ≥47.5% (Theory 63.08%, dry wt. basis)
Loss on Drying≤25% (200°C/constant wt.)
12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

Solubility
Soluble in water, ethanol and ether.

Notes
Hygroscopic. Incompatible with organic materials and powdered metals.
RUO – Research Use Only

General References:

  1. Sharma, P.; Patel, A. Zirconia supported 12-molybdophosphoric acid: Physico-chemical characterization and non-solvent liquid phase oxidation of styrene. Appl. Surf. Sci. 2009, 55 (17), 7635-7641.
  2. Fan, D.; Li, G.; Hao, J. Fabrication and electrocatalytic activities of porphyrin and 12-molybdophosphoric acid hybrid films. J. Colloid Interface Sci. 2010, 351 (1), 151-155.