Zinc trifluoromethanesulfonate, 98%
Zinc trifluoromethanesulfonate, 98%
Zinc trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Zinc trifluoromethanesulfonate, 98%

CAS: 54010-75-2 | C2F6O6S2Zn | 363.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L15969.06
also known as L15969-06
Price (USD)
57.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
57.40
Each
Chemical Identifiers
CAS54010-75-2
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream
Assay (unspecified)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.0%
Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

Solubility
Soluble in water and acetonitrile. Slightly soluble in methanol. Insoluble in dichloromethane.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. Mild Lewis acid catalyst which promotes thioketalization, particularly of acid-sensitive substrates, with ethanedithiol: Tetrahedron Lett., 24, 169 (1983); J. Am. Chem. Soc., 105, 1662 (1983).
  2. Useful, alone or in combination with TMS chloride, for glycosidation reactions: Chem. Pharm. Bull., 40, 2909 (1992); 41, 201 (1993); 42, 1808 (1994).
  3. Promotes the aldol reaction of silyl enol ethers with aldehydes: Gazz. Chim. Ital., 123, 673 (1993).
  4. In combination with N-methylephedrine, catalyzes enantioselective addition of terminal alkynes to aldehydes to give propargylic alcohols in high yield and ee: J. Am. Chem. Soc., 122, 1806 (2000).
  5. Castro-Osma, J. A.; Comerford, J. W.; Heyn, R. H.; North, M.; Tangstad, E. New catalysts for carboxylation of propylene glycol to propylene carbonate via high-throughput screening. Faraday Discuss. 2015, 183, 19-30.
  6. Sownthari, K.; Suthanthiraraj, S. A. Influence of Al2O3 nanofiller on the properties of polymer electrolyte based on poly-epsilon-caprolactone. Polym. Bull. 2015, 72 (1), 61-73.