4-Iodophenyl isocyanate, 97%
4-Iodophenyl isocyanate, 97%
4-Iodophenyl isocyanate, 97%
4-Iodophenyl isocyanate, 97%
Thermo Scientific Chemicals

4-Iodophenyl isocyanate, 97%

CAS: 15845-62-2 | C7H4INO | 245.019 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L16000.03
also known as L16000-03
Price (USD)
43.65
Online Exclusive
48.60
Save 4.95 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.60
Save 4.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS15845-62-2
IUPAC Name1-iodo-4-isocyanatobenzene
Molecular FormulaC7H4INO
InChI KeyKVMKUTIUJGYHBA-UHFFFAOYSA-N
SMILESIC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange to brown
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥96.0%
Melting Point (clear melt)41.0-50.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only