2-Methoxy-5-methylphenyl isocyanate, 97%
2-Methoxy-5-methylphenyl isocyanate, 97%
2-Methoxy-5-methylphenyl isocyanate, 97%
2-Methoxy-5-methylphenyl isocyanate, 97%
Thermo Scientific Chemicals

2-Methoxy-5-methylphenyl isocyanate, 97%

CAS: 59741-04-7 | C9H9NO2 | 163.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L16001.06
also known as L16001-06
Price (USD)
62.65
Online Exclusive
69.40
Save 6.75 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
69.40
Save 6.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS59741-04-7
IUPAC Name2-isocyanato-1-methoxy-4-methylbenzene
Molecular FormulaC9H9NO2
InChI KeyIDOHLSFNBKNRRJ-UHFFFAOYSA-N
SMILESCOC1=C(C=C(C)C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5335-1.5385 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only