Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)
Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)
Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)
Thermo Scientific Chemicals

Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)

CAS: 6074-84-6 | C10H25O5Ta | 406.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L16033.06
also known as L16033-06
Price (USD)
94.65
Special offer
Online exclusive
Ends: 30-Jun-2026
111.00
Save 16.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
94.65
Special offer
Online exclusive
Ends: 30-Jun-2026
111.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS6074-84-6
IUPAC Namepentaethoxytantalum
Molecular FormulaC10H25O5Ta
InChI KeyHSXKFDGTKKAEHL-UHFFFAOYSA-N
SMILESCCO[Ta](OCC)(OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid, may contain crystalline solid on standing
Appearance (Color)Clear colorless to pale yellow
Total Metal Impurities≤500ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only