6-Bromo-2-naphthoic acid, 98%
6-Bromo-2-naphthoic acid, 98%
6-Bromo-2-naphthoic acid, 98%
Thermo Scientific Chemicals

6-Bromo-2-naphthoic acid, 98%

CAS: 5773-80-8 | C11H7BrO2 | 251.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16093.06
also known as L16093-06
Price (USD)
50.65
Online exclusive
55.80
Save 5.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.65
Online exclusive
55.80
Save 5.15
Each
Chemical Identifiers
CAS5773-80-8
IUPAC Name6-bromonaphthalene-2-carboxylic acid
Molecular FormulaC11H7BrO2
InChI KeyNPMCAVBMOTZUPD-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2C=C(Br)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormPowder or lumps
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. V. Krishnakumar,; R. Mathammal,; S. Muthunatesan. Structures and vibrational frequencies of 2-naphthoic acid and 6-bromo-2-naphthoic acid based on density functional theory calculations. JouSpectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2008, 70(1),201-209.
  2. G. B. Patel,; R. R. Mahire,; D. H. More. Synthesis and characterization of benzimidazole derivatives by sonogashira coupling and evaluation of anthelmintic activity. Der Pharma Chemica,. year of publication , 5 (6),369-376 .