(R)-(+)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%
(R)-(+)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%
(R)-(+)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%
Thermo Scientific Chemicals

(R)-(+)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%

CAS: 88196-70-7 | C9H13NO | 151.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16323.14
also known as L16323-14
Price (USD)
451.65
Online Exclusive
502.00
Save 50.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
451.65
Online Exclusive
502.00
Save 50.35 (10%)
Each
Chemical Identifiers
CAS88196-70-7
IUPAC Name1-(3-methoxyphenyl)ethan-1-amine
Molecular FormulaC9H13NO
InChI KeyCJWGCBRQAHCVHW-UHFFFAOYNA-N
SMILESCOC1=CC=CC(=C1)C(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
Enantiomeric excess≥97.5%
Optical Rotation+27? ±1? (Neat)
View more
(R)-(+)-1-(3-Methoxyphenyl)ethylamine is used in dietary supplements and cosmeticeuticals. It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-(3-Methoxyphenyl)ethylamine is used in dietary supplements and cosmeticeuticals. It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (10g/L at 20°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. It is sensitive to air. Store under dry inert gas. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Kenichi Sakai.; Yukihiko Hashimoto.; Kazushi Kinbara.; Kazuhiko Saigo.; Hisamichi Murakami.; Hiroyuki Nohira. Optical Resolution of 1-(3-Methoxyphenyl)ethylamine with Enantiomerically Pure Mandelic Acid, and the Crystal Structure of Less-Soluble Diastereomeric Salt. Bulletin of the Chemical Society of Japan. 1993, 66 (11), 3414-3418.
  2. Hu, Meng.; Zhang, Fu-Li.; Xie, Mei-Hua. A Simple and Efficient Synthesis of (S)- and (R)-1-(3-Methoxyphenyl) Ethylamine. Letters in Organic Chemistry. 2007, 4 (2), 126-128.