(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%
(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%
(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%
Thermo Scientific Chemicals

(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%

CAS: 82796-69-8 | C9H13NO | 151.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L16324.14
also known as L16324-14
Price (USD)
384.65
Online Exclusive
427.00
Save 42.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
384.65
Online Exclusive
427.00
Save 42.35 (10%)
Each
Chemical Identifiers
CAS82796-69-8
IUPAC Name1-(3-methoxyphenyl)ethan-1-amine
Molecular FormulaC9H13NO
InChI KeyCJWGCBRQAHCVHW-UHFFFAOYNA-N
SMILESCOC1=CC=CC(=C1)C(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-27° ± 2°(neat)
Appearance (Color)Clear colourless to very pale yellow
Enantiomeric excess>99.0% by GC
Assay (GC)≥99.0%
Refractive Index1.5325-1.5345 @ 20°C
View more
Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

Solubility
Soluble in water (10g/L).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only
Hu, Meng; Zhang, Fu-Li; Xie, Mei-Hua. A Simple and Efficient Synthesis of (S)- and (R)-1-(3-Methoxyphenyl) Ethylamine. Letters in Organic Chemistry. 2007, 4(2), 126-128(3).