(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%
(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%
(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%
(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%
Thermo Scientific Chemicals

(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%

CAS: 27298-98-2 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L16329.14
also known as L16329-14
Price (USD)
272.65
Special offer
Online exclusive
Ends: 30-Jun-2026
321.00
Save 48.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
272.65
Special offer
Online exclusive
Ends: 30-Jun-2026
321.00
Save 48.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS27298-98-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless
Formliquid
Assay (GC)> 97.5%
Enantiomeric excess> 99.0% by GC
Optical Rotation-36? ? 2? (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only