Ethyl 3-methylisoxazole-4-carboxylate, 97%
Ethyl 3-methylisoxazole-4-carboxylate, 97%
Ethyl 3-methylisoxazole-4-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 3-methylisoxazole-4-carboxylate, 97%

CAS: 20328-15-8 | C7H9NO3 | 155.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L16362.03
also known as L16362-03
Price (USD)
37.65
Online Exclusive
41.50
Save 3.85 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.50
Save 3.85 (9%)
Each
Chemical Identifiers
CAS20328-15-8
IUPAC Nameethyl 3-methyl-1,2-oxazole-4-carboxylate
Molecular FormulaC7H9NO3
InChI KeyAEWHICNMVQOWJE-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CON=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥96.0%
Refractive Index1.4570-1.4620 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only