5-Methylnicotinic acid, 97%
5-Methylnicotinic acid, 97%
5-Methylnicotinic acid, 97%
5-Methylnicotinic acid, 97%
Thermo Scientific Chemicals

5-Methylnicotinic acid, 97%

CAS: 3222-49-9 | C7H7NO2 | 137.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L16370.03
also known as L16370-03
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS3222-49-9
IUPAC Name5-methylpyridine-3-carboxylic acid
Molecular FormulaC7H7NO2
InChI KeyDJDHHXDFKSLEQY-UHFFFAOYSA-N
SMILESCC1=CN=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Melting Point (clear melt)208.0-218.0?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Sparingly soluble (0.083 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only