Cyclopentyltrimethoxysilane, 95%
Cyclopentyltrimethoxysilane, 95%
Cyclopentyltrimethoxysilane, 95%
Thermo Scientific Chemicals

Cyclopentyltrimethoxysilane, 95%

CAS: 143487-47-2 | C8H18O3Si | 190.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16442.14
also known as L16442-14
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.60
Save 13.95 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.60
Save 13.95 (15%)
Each
Chemical Identifiers
CAS143487-47-2
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥94.0%
FormLiquid
Appearance (Color)Clear, colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only