Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%
Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%
Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%
Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%
Thermo Scientific Chemicals

Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%

CAS: 2062-98-8 | C6F12O2 | 332.045 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L16483.14
also known as L16483-14
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
314.65
Online Exclusive
349.00
Save 34.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2062-98-8
IUPAC Name2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoyl fluoride
Molecular FormulaC6F12O2
InChI KeyBCLQALQSEBVVAD-UHFFFAOYNA-N
SMILESFC(=O)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index<1.3000 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only