5-Fluorocytosine, 98+%
5-Fluorocytosine, 98+%
5-Fluorocytosine, 98+%
Thermo Scientific Chemicals

5-Fluorocytosine, 98+%

CAS: 2022-85-7 | C4H4FN3O | 129.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L16496.03
also known as L16496-03
Price (USD)
92.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
92.60
Each
Chemical Identifiers
CAS2022-85-7
IUPAC Name6-amino-5-fluoro-1,2-dihydropyrimidin-2-one
Molecular FormulaC4H4FN3O
InChI KeyXRECTZIEBJDKEO-UHFFFAOYSA-N
SMILESNC1=C(F)C=NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formcrystalline powder
Assay (HPLC)>98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
5-Fluorocytosine acts as an antidiabetic, antifungal and antimicrobial agent. It is useful for the treatment of serious infections arises due to susceptible strains of Candida or Cryptococcus neoformans and chromomycosis. Further, it is employed in studies on TMP biosynthesis.

Solubility
Soluble in water and hydrochloric acid.

Notes
Light Sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Olszewska, A.; Dadová, J.; Mačková, M.; Hocek, M. Cleavage of DNA containing 5-fluorocytosine or 5-fluorouracil by type II restriction endonucleases. Bioorg. Med. Chem. 2015, 23 (21), 6885-6890.
  2. Perumalla, S. R.; Sun, C. C. Enabling Tablet Product Development of 5-Fluorocytosine Through Integrated Crystal and Particle Engineering. J. Pharm. Sci. 2014, 103 (4), 1126-1132.