Octamethylcyclotetrasilazane, 97%
Octamethylcyclotetrasilazane, 97%
Octamethylcyclotetrasilazane, 97%
Thermo Scientific Chemicals

Octamethylcyclotetrasilazane, 97%

CAS: 1020-84-4 | C8H28N4Si4 | 292.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16563.14
also known as L16563-14
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.50
Save 10.85 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.50
Save 10.85 (15%)
Each
Chemical Identifiers
CAS1020-84-4
IUPAC Name2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
Molecular FormulaC8H28N4Si4
InChI KeyFIADVASZMLCQIF-UHFFFAOYSA-N
SMILESC[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)93.5-99.5?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only