2,2,3,3-Tetrafluoro-1,4-butanediol, 97%
2,2,3,3-Tetrafluoro-1,4-butanediol, 97%
2,2,3,3-Tetrafluoro-1,4-butanediol, 97%
2,2,3,3-Tetrafluoro-1,4-butanediol, 97%
Thermo Scientific Chemicals

2,2,3,3-Tetrafluoro-1,4-butanediol, 97%

CAS: 425-61-6 | C4H6F4O2 | 162.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16635.06
also known as L16635-06
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.20
Save 14.55 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
95.20
Save 14.55 (15%)
Each
Add to cart
Chemical Identifiers
CAS425-61-6
IUPAC Name2,2,3,3-tetrafluorobutane-1,4-diol
Molecular FormulaC4H6F4O2
InChI KeyCDZXJJOGDCLNKX-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Melting Point (clear melt)78.0-88.0?C
Assay (Silylated GC)≥96.0%
2,2,3,3-Tetrafluoro-1,4-butanediol is a reactant used for the enzymic synthesis of silicone fluorinated aliphatic polyester amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluoro-1,4-butanediol is a reactant used for the enzymic synthesis of silicone fluorinated aliphatic polyester amides.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
RUO – Research Use Only

General References:

  1. Palsule, A. S.; Poojari, Y. Enzymatic synthesis of silicone fluorinated aliphatic polyester amides. Polymer. 2010, 51 6161-6167.
  2. Ashish Vaidya.; Manoj K. Chaudhury. Synthesis and Surface Properties of Environmentally Responsive Segmented Polyurethanes. Journal of Colloid and Interface Science Chemistry. 2002, 249 (1), 235-245.