Tetra-n-propoxysilane, 97%
Tetra-n-propoxysilane, 97%
Tetra-n-propoxysilane, 97%
Thermo Scientific Chemicals

Tetra-n-propoxysilane, 97%

CAS: 682-01-9 | C12H28O4Si | 264.437 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16644.22
also known as L16644-22
Price (USD)
84.65
Online Exclusive
93.50
Save 8.85 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
93.50
Save 8.85 (9%)
Each
Chemical Identifiers
CAS682-01-9
IUPAC Nametetrapropyl silicate
Molecular FormulaC12H28O4Si
InChI KeyZQZCOBSUOFHDEE-UHFFFAOYSA-N
SMILESCCCO[Si](OCCC)(OCCC)OCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3990-1.4030 @ 20°C (non-U.S. specification)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only