Ethoxytriethylsilane, 97%
Ethoxytriethylsilane, 97%
Ethoxytriethylsilane, 97%
Ethoxytriethylsilane, 97%
Thermo Scientific Chemicals

Ethoxytriethylsilane, 97%

CAS: 597-67-1 | C8H20OSi | 160.332 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16656.14
also known as L16656-14
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS597-67-1
IUPAC Nameethoxytriethylsilane
Molecular FormulaC8H20OSi
InChI KeyDFJDZTPFNSXNAX-UHFFFAOYSA-N
SMILESCCO[Si](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4115-1.4165 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only