Dimethylethoxyvinylsilane, 97%
Dimethylethoxyvinylsilane, 97%
Dimethylethoxyvinylsilane, 97%
Thermo Scientific Chemicals

Dimethylethoxyvinylsilane, 97%

CAS: 5356-83-2 | C6H14OSi | 130.262 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16699.22
also known as L16699-22
Price (USD)
275.65
Online Exclusive
306.00
Save 30.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
275.65
Online Exclusive
306.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS5356-83-2
IUPAC Nameethenyl(ethoxy)dimethylsilane
Molecular FormulaC6H14OSi
InChI KeyJEWCZPTVOYXPGG-UHFFFAOYSA-N
SMILESCCO[Si](C)(C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.3955-1.4005 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only