2-Bromo-4,5-difluorophenol, 97%
2-Bromo-4,5-difluorophenol, 97%
2-Bromo-4,5-difluorophenol, 97%
Thermo Scientific Chemicals

2-Bromo-4,5-difluorophenol, 97%

CAS: 166281-37-4 | C6H3BrF2O | 208.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L16716.03
also known as L16716-03
Price (USD)
34.65
Online Exclusive
38.50
Save 3.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
38.50
Save 3.85 (10%)
Each
Chemical Identifiers
CAS166281-37-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to yellow to gray
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5290-1.5340 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only