1H,1H,2H-Perfluoro-1-hexene, 97%
1H,1H,2H-Perfluoro-1-hexene, 97%
1H,1H,2H-Perfluoro-1-hexene, 97%
1H,1H,2H-Perfluoro-1-hexene, 97%
Thermo Scientific Chemicals

1H,1H,2H-Perfluoro-1-hexene, 97%

CAS: 19430-93-4 | C6H3F9 | 246.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L16834.14
also known as L16834-14
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS19430-93-4
IUPAC Name3,3,4,4,5,5,6,6,6-nonafluorohex-1-ene
Molecular FormulaC6H3F9
InChI KeyGVEUEBXMTMZVSD-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)>96.0%
Refractive Index<1.3000 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only