Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene)
Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene)
Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene)
Thermo Scientific Chemicals

Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene)

CAS: 2070-70-4 | C6F12 | 300.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16836.14
also known as L16836-14
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.90
Save 8.25 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 30-Jun-2026
53.90
Save 8.25 (15%)
Each
Chemical Identifiers
CAS2070-70-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)>90.0% (main component)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only