4,4,4-Trifluorocrotononitrile, 96%
4,4,4-Trifluorocrotononitrile, 96%
4,4,4-Trifluorocrotononitrile, 96%
Thermo Scientific Chemicals

4,4,4-Trifluorocrotononitrile, 96%

CAS: 406-86-0 | C4H2F3N | 121.06 g/mol
Have Questions?
Catalog NumberQuantity
L16870.03
also known as L16870-03
1 g
Catalog number L16870.03
also known as L16870-03
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
107.00
Save 16.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
107.00
Save 16.35
Each
Chemical Identifiers
CAS406-86-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.3405-1.3465 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only