2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std.
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std.
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std.
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std.
Thermo Scientific Chemicals

2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std.

CAS: 915-76-4 | C12F21N3 | 585.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L16883.03
also known as L16883-03
Price (USD)
81.90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
81.90
Each
Add to cart
Specifications
Beilstein381010
CAS915-76-4
Chemical Name or Material2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
Density1.716
EINECS Number213-023-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only