Hexamethyldisiloxane, NMR grade, 99.7%
Hexamethyldisiloxane, NMR grade, 99.7%
Hexamethyldisiloxane, NMR grade, 99.7%
Thermo Scientific Chemicals

Hexamethyldisiloxane, NMR grade, 99.7%

CAS: 107-46-0 | C6H18OSi2 | 162.379 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16970.22
also known as L16970-22
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
67.10
Save 9.45 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
67.10
Save 9.45 (14%)
Each
Chemical Identifiers
CAS107-46-0
IUPAC Nametrimethyl[(trimethylsilyl)oxy]silane
Molecular FormulaC6H18OSi2
InChI KeyUQEAIHBTYFGYIE-UHFFFAOYSA-N
SMILESC[Si](C)(C)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥99.5% (GC, US sourced material)
CommentMaterial sourced in UK or US
Identification (FTIR)Conforms (UK sourced material)
Refractive Index1.3755-1.3785 @ 20?C (UK sourced material)
View more
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
RUO – Research Use Only

General References:

  1. Bashir, M.; Bashir, S. Hydrophobic-Hydrophilic Character of Hexamethyldisiloxane Films Polymerized by Atmospheric Pressure Plasma Jet. Plasma Chem. Plasma Process 2015, 35 (4), 739-755.
  2. Hsiao, C. R.; Lin, C. W.; Chou, C. M.; Chung, C. J.; He, J. L. Surface modification of blood-contacting biomaterials by plasma-polymerized superhydrophobic films using hexamethyldisiloxane and tetrafluoromethane as precursors. Appl. Surf. Sci. 2015, 346, 50-56.
  3. Ershov, S.; Khelifa, F.; Druart, M. E.; Habibi, Y.; Olivier, M. G.; Snyders, R.; Dubois, P. Free radical-induced grafting from plasma polymers for the synthesis of thin barrier coatings. RSC Adv. 2015, 5 (19), 14256-14265.
  4. Khelifa, F.; Ershov, S.; Druart, M. E.; Habibi, Y.; Chicot, D.; Olivier, M. G.; Snydersa, R.; Dubois, P. A multilayer coating with optimized properties for corrosion protection of Al. J. Mater. Chem. A 2015, 3 (31), 15977-15985.