(1S,2S)-(+)-2-Benzyloxycyclohexylamine, ChiPros 99+%, ee 99%
(1S,2S)-(+)-2-Benzyloxycyclohexylamine, ChiPros 99+%, ee 99%
(1S,2S)-(+)-2-Benzyloxycyclohexylamine, ChiPros 99+%, ee 99%
Thermo Scientific Chemicals

(1S,2S)-(+)-2-Benzyloxycyclohexylamine, ChiPros 99+%, ee 99%

CAS: 216394-07-9 | C13H19NO | 205.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17020.03
also known as L17020-03
Price (USD)
53.65
Online Exclusive
59.50
Save 5.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.50
Save 5.85 (10%)
Each
Chemical Identifiers
CAS216394-07-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Formliquid
Assay (GC)> 99.0%
Enantiomeric excess≥ 99% by GC
Refractive Index1.5265 - 1.5295 @20?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only