Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%
Thermo Scientific Chemicals

Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%

CAS: 72022-68-5 | C22H40N2S10Zn | 718.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L17046.MD
also known as L17046-MD
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 30-Jun-2026
102.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS72022-68-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Bright red
FormPowder
Elemental AnalysisCarbon: 35.30-38.24%
Elemental AnalysisNitrogen: 3.74-4.06%
Elemental AnalysisSulfur: 42.83-46.40%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only