2-Chloro-6-nitrobenzoic acid, 99%
2-Chloro-6-nitrobenzoic acid, 99%
2-Chloro-6-nitrobenzoic acid, 99%
Thermo Scientific Chemicals

2-Chloro-6-nitrobenzoic acid, 99%

CAS: 5344-49-0 | C7H4ClNO4 | 201.562 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17059.06
also known as L17059-06
Price (USD)
319.65
Online Exclusive
355.00
Save 35.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
319.65
Online Exclusive
355.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS5344-49-0
IUPAC Name2-chloro-6-nitrobenzoic acid
Molecular FormulaC7H4ClNO4
InChI KeyJYHOMEFOTKWQPN-UHFFFAOYSA-N
SMILESOC(=O)C1=C(Cl)C=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to orange to brown
FormCrystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only