(R)-(+)-2-Benzyloxypropionic acid, 98%
(R)-(+)-2-Benzyloxypropionic acid, 98%
(R)-(+)-2-Benzyloxypropionic acid, 98%
Thermo Scientific Chemicals

(R)-(+)-2-Benzyloxypropionic acid, 98%

CAS: 100836-85-9 | C10H12O3 | 180.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L17205.22
also known as L17205-22
Price (USD)
385.65
Special offer
Online exclusive
Ends: 30-Jun-2026
453.00
Save 67.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
385.65
Special offer
Online exclusive
Ends: 30-Jun-2026
453.00
Save 67.35 (15%)
Each
Chemical Identifiers
CAS100836-85-9
IUPAC Name2-(benzyloxy)propanoic acid
Molecular FormulaC10H12O3
InChI KeyXWAVPOFYNPXXEL-UHFFFAOYNA-N
SMILESCC(OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Silylated GC)≥97.5%
Optical Rotation+77± 3?, c = 1% in Methanol
FormCrystals or powder or crystalline powder
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
Yoshiko Tsuda.; Kenichi Kawai.; Shoichi Nakajima. Microbial Reduction of 2-Phenylpropionic Acid, 2-Benzyloxypropionic Acid and 2-(2-Furfuryl) propionic Acid . Chemical and Pharmaceutical Bulletin. 1985, 33 (11), 4657-4661.