10,11-Dihydrocarbamazepine, 99%
10,11-Dihydrocarbamazepine, 99%
10,11-Dihydrocarbamazepine, 99%
Thermo Scientific Chemicals

10,11-Dihydrocarbamazepine, 99%

CAS: 3564-73-6 | C15H14N2O | 238.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Catalog number L17271.MF
also known as L17271-MF
Price (USD)
53.70
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
53.70
Each
Chemical Identifiers
CAS3564-73-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to cream to pale yellow
Assay (HPLC)≥98.5%
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.

Solubility
Soluble in dichloromethane. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. M.M Bhatti.; G.D Hanson.; L Schultz. Simultaneous determination of phenytoin, carbamazepine, and 10,11-carbamazepine epoxide in human plasma by high-performance liquid chromatography with ultraviolet detection1. Journal of Pharmaceutical and Biomedical Analysis. 1998, 16 (7), 1233-1240.
  2. Michel Eichelbaum.; Leif Bertilsson. Determination of carbamazepine and its epoxide metabolite in plasma by high-speed liquid chromatography. Journal of Chromatography A. 1975, 103 (1), 135-140.