n-Dodecyltriethoxysilane, 95%
n-Dodecyltriethoxysilane, 95%
n-Dodecyltriethoxysilane, 95%
Thermo Scientific Chemicals

n-Dodecyltriethoxysilane, 95%

CAS: 18536-91-9 | C18H40O3Si | 332.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L17293.14
also known as L17293-14
Price (USD)
83.65
Online Exclusive
92.30
Save 8.65 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.30
Save 8.65 (9%)
Each
Chemical Identifiers
CAS18536-91-9
IUPAC Namedodecyltriethoxysilane
Molecular FormulaC18H40O3Si
InChI KeyYGUFXEJWPRRAEK-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Refractive Index1.4235-1.4295 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only