3,5-Difluorobenzeneboronic acid, 97+%
3,5-Difluorobenzeneboronic acid, 97+%
3,5-Difluorobenzeneboronic acid, 97+%
Thermo Scientific Chemicals

3,5-Difluorobenzeneboronic acid, 97+%

CAS: 156545-07-2 | C6H5BF2O2 | 157.911 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17425.03
also known as L17425-03
Price (USD)
26.65
Online exclusive
29.50
Save 2.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
26.65
Online exclusive
29.50
Save 2.85
Each
Chemical Identifiers
CAS156545-07-2
IUPAC Name(3,5-difluorophenyl)boronic acid
Molecular FormulaC6H5BF2O2
InChI KeyQWQBQRYFWNIDOC-UHFFFAOYSA-N
SMILESOB(O)C1=CC(F)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.0%
Assay (Aqueous acid-base Titration)≥97.0%
Proton NMRConforms to structure
FormPowder
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only