1-Bromo-3-(tert-butyldimethylsiloxy)benzene, 98+%, Thermo Scientific Chemicals
1-Bromo-3-(tert-butyldimethylsiloxy)benzene, 98+%, Thermo Scientific Chemicals
1-Bromo-3-(tert-butyldimethylsiloxy)benzene, 98+%, Thermo Scientific Chemicals
1-Bromo-3-(tert-butyldimethylsiloxy)benzene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-3-(tert-butyldimethylsiloxy)benzene, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L17440.06
also known as L17440-06
5 g
Catalog number L17440.06
also known as L17440-06
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Add to cart
Chemical Identifiers
CAS65423-56-5
IUPAC Name(3-bromophenoxy)(tert-butyl)dimethylsilane
Molecular FormulaC12H19BrOSi
InChI KeyBFRHMVJJJGUHDI-UHFFFAOYSA-N
SMILESCC(C)(C)[Si](C)(C)OC1=CC(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5070-1.5110 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only