5-Isopropyl-2-methoxybenzeneboronic acid, 98+%
5-Isopropyl-2-methoxybenzeneboronic acid, 98+%
5-Isopropyl-2-methoxybenzeneboronic acid, 98+%
Thermo Scientific Chemicals

5-Isopropyl-2-methoxybenzeneboronic acid, 98+%

CAS: 216393-63-4 | C10H15BO3 | 194.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17460.03
also known as L17460-03
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.80
Save 8.15
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.80
Save 8.15
Each
Chemical Identifiers
CAS216393-63-4
IUPAC Name[2-methoxy-5-(propan-2-yl)phenyl]boronic acid
Molecular FormulaC10H15BO3
InChI KeyUTKAEAGLVJFBMK-UHFFFAOYSA-N
SMILESCOC1=C(C=C(C=C1)C(C)C)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥98.0%
Assay (HPLC)≥98.0%
Melting Point (clear melt)81.0-87.0?C
Proton NMRConforms to structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only