5-Fluoroindole-3-carboxaldehyde, 98%
5-Fluoroindole-3-carboxaldehyde, 98%
5-Fluoroindole-3-carboxaldehyde, 98%
Thermo Scientific Chemicals

5-Fluoroindole-3-carboxaldehyde, 98%

CAS: 2338-71-8 | C9H6FNO | 163.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L17507.06
also known as L17507-06
Price (USD)
518.65
Online Exclusive
576.00
Save 57.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
518.65
Online Exclusive
576.00
Save 57.35 (10%)
Each
Chemical Identifiers
CAS2338-71-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to pale orange
FormPowder
Assay (GC)≥97.5%
Free acid (titration)≤1.0%
Melting Point (clear melt)164.0-170.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only