2,3,5-Trichlorobenzeneboronic acid, 98%
2,3,5-Trichlorobenzeneboronic acid, 98%
2,3,5-Trichlorobenzeneboronic acid, 98%
2,3,5-Trichlorobenzeneboronic acid, 98%
Thermo Scientific Chemicals

2,3,5-Trichlorobenzeneboronic acid, 98%

CAS: 212779-19-6 | C6H4BCl3O2 | 225.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17511.03
also known as L17511-03
Price (USD)
62.50
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
62.50
Each
Add to cart
Chemical Identifiers
CAS212779-19-6
IUPAC Name(2,3,5-trichlorophenyl)boronic acid
Molecular FormulaC6H4BCl3O2
InChI KeyOPBCCRZCYTUJMS-UHFFFAOYSA-N
SMILESOB(O)C1=CC(Cl)=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormPowder or lumps or fused solid
Assay (Aqueous acid-base Titration)≥97.5%
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only