2-Amino-5-chlorobenzaldehyde, 97%
2-Amino-5-chlorobenzaldehyde, 97%
2-Amino-5-chlorobenzaldehyde, 97%
2-Amino-5-chlorobenzaldehyde, 97%
Thermo Scientific Chemicals

2-Amino-5-chlorobenzaldehyde, 97%

CAS: 20028-53-9 | C7H6ClNO | 155.581 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17515.03
also known as L17515-03
Price (USD)
72.65
Online Exclusive
80.60
Save 7.95 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.60
Save 7.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS20028-53-9
IUPAC Name2-amino-5-chlorobenzaldehyde
Molecular FormulaC7H6ClNO
InChI KeyKUIFMGITZFDQMP-UHFFFAOYSA-N
SMILESNC1=C(C=O)C=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to yellow to orange to brown
FormCrystals or powder or crystalline powder or fused solid or granules
Assay (HPLC)≥96.0%
Free acid (titration)≤1.5%
Melting Point (clear melt)67.0-76.0?C
2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Smuszkiewicz, A.; López-Sanz, J.; Pérez-Mayoral, E.; Soriano, E.; Sobczak, I.; Ziolek, M. ; Martín-Aranda, R. M.; López-Peinado, A. J. Amino-grafted mesoporous materials based on MCF structure involved in the quinoline synthesis. Mechanistic insights. J. Mol. Catal. A: Chem. 2013, 378, 38-46.
  2. Smuszkiewicz, A.; Pérez-Mayoral, E.; Soriano, E.; Sobczak, I.; Ziolek, M.; Martín-Aranda, R. M.; López-Peinado, A. J. Bifunctional mesoporous MCF materials as catalysts in the Friedländer condensation. Catal. Today 2013, 218-219, 70-75.
  3. Nammalwar, B.; Murie, M.; Fortenberry, C.; Bunce,R. A. Quinoline- and 1,8-naphthyridine-3-carboxylic acids using a self-catalyzed Friedländer approach. Tetrahedron Lett. 2014, 55 (20), 3181-3183.
  4. Gouda, M. A.; Sabah, M. A.; Aljuhani, W. K.; El-Gahani, A. S.; El-Enazi, S. A. E.; Al Enizi, S. A.; Al-Balawi, M. M. Utility of 2-cyano-N-(2-hydroxyethyl) acetamide in heterocyclic synthesis. Eur. J. Chem.2015, 6 (2), 219-224.