5-Methyl-2-hexanone, 99%
5-Methyl-2-hexanone, 99%
5-Methyl-2-hexanone, 99%
Thermo Scientific Chemicals

5-Methyl-2-hexanone, 99%

CAS: 110-12-3 | C7H14O | 114.188 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number L17584.AE
also known as L17584-AE
Price (USD)
23.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.00
Save 4.35 (16%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.00
Save 4.35 (16%)
Each
Chemical Identifiers
CAS110-12-3
IUPAC Name5-methylhexan-2-one
Molecular FormulaC7H14O
InChI KeyFFWSICBKRCICMR-UHFFFAOYSA-N
SMILESCC(C)CCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4060-1.4090 @ 20°C
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (5.4 g/L)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Nien-Chu Yang.; Steven P. Elliott. Photochemistry of (s)-(+)-5-methyl-2-heptanone.J. Am. Chem. Soc. 1969, 91 (26), 7550-7551 .
  2. S.B. Jadhav.; S.P. Tandel.; S.P. Malve. Extraction and spectrophotometric determination of cobalt(II) with isonitroso-5-methyl-2-hexanone.Talanta. 2001, 55 (6), 1059-1064 .