7-Chloroindole, 98%
7-Chloroindole, 98%
7-Chloroindole, 98%
Thermo Scientific Chemicals

7-Chloroindole, 98%

CAS: 53924-05-3 | C8H6ClN | 151.593 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L17637.03
also known as L17637-03
Price (USD)
87.65
Special offer
Online exclusive
Ends: 30-Jun-2026
103.00
Save 15.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
87.65
Special offer
Online exclusive
Ends: 30-Jun-2026
103.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS53924-05-3
IUPAC Name7-chloro-1H-indole
Molecular FormulaC8H6ClN
InChI KeyWMYQAKANKREQLM-UHFFFAOYSA-N
SMILESClC1=C2NC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder or powder
Assay (GC)≥97.5%
Melting point55-60°C
Appearance (Color)White to cream to yellow to pale brown to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only