2-Amino-5-bromo-3,4-dimethylpyridine, 97%
2-Amino-5-bromo-3,4-dimethylpyridine, 97%
2-Amino-5-bromo-3,4-dimethylpyridine, 97%
Thermo Scientific Chemicals

2-Amino-5-bromo-3,4-dimethylpyridine, 97%

CAS: 374537-97-0 | C7H9BrN2 | 201.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17672.06
also known as L17672-06
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
311.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS374537-97-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown
Assay (GC)≥96.0%
Melting Point (clear melt)129.0-138.0?C
FormCrystals or powder or crystalline powder
It is an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Evgeny Kiselev.; Keli Agama.; Yves Pommier.; Mark Cushman. Azaindenoisoquinolines as Topoisomerase I Inhibitors and Potential Anticancer Agents: A Systematic Study of Structure-Activity Relationships.J. Med. Chem. 2012, 55 (4),1682-1697 .