3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%
3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%
3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%
3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%

CAS: 33558-36-0 | C10H9F6Si | 271.257 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17673.03
also known as L17673-03
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.40
Save 8.75 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.40
Save 8.75 (14%)
Each
Add to cart
Chemical Identifiers
CAS33558-36-0
IUPAC Name[3,5-bis(trifluoromethyl)phenyl]dimethylsilyl
Molecular FormulaC10H9F6Si
InChI KeyHJQABLGRCWBCDT-UHFFFAOYSA-N
SMILESC[Si](C)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4100-1.4160 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
3,5-Bis(trifluoromethyl)phenyldimethylsilane is used as the Intermediates of organic light-emitting diode and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)phenyldimethylsilane is used as the Intermediates of organic light-emitting diode and as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only