Dimethyl naphthalene-2,6-dicarboxylate, 99+%
Dimethyl naphthalene-2,6-dicarboxylate, 99+%
Dimethyl naphthalene-2,6-dicarboxylate, 99+%
Thermo Scientific Chemicals

Dimethyl naphthalene-2,6-dicarboxylate, 99+%

CAS: 840-65-3 | C14H12O4 | 244.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L17689.06
also known as L17689-06
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.60
Save 14.95
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.60
Save 14.95
Each
Chemical Identifiers
CAS840-65-3
IUPAC Name2,6-dimethyl naphthalene-2,6-dicarboxylate
Molecular FormulaC14H12O4
InChI KeyGYUVMLBYMPKZAZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC2=CC=C(C=C2C=C1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)188-198?C
Appearance (Color)White to pale cream or pale yellow
Assay (chromatographic, GC or HPLC)≥99.0%
FormPowder or crystals or crystalline powder or flakes
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

Solubility
Soluble in hot toluene (very faint turbidity). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. George P. Karayannidis.; George Z. Papageorgiou.; Demetris N. Bikiaris.; Eleftherios V. Tourasanidis. Synthesis and thermal behaviour of poly(ethylene-co-butylene naphthalene-2,6-dicarboxylate)s. Langmuir. 1998, 39 (17), 4129-4134.
  2. H. Zhang.; I. M. Ward. Kinetics of Hydrolytic Degradation of Poly(ethylene naphthalene-2,6-dicarboxylate). Langmuir. 1995, 28 (23), 7622-7629.