4-(Methylsulfinyl)benzeneboronic acid, 98%
4-(Methylsulfinyl)benzeneboronic acid, 98%
4-(Methylsulfinyl)benzeneboronic acid, 98%
Thermo Scientific Chemicals

4-(Methylsulfinyl)benzeneboronic acid, 98%

CAS: 166386-48-7 | C7H9BO3S | 184.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number L17865.03
also known as L17865-03
Price (USD)
203.65
Special offer
Online exclusive
Ends: 28-Dec-2026
239.00
Save 35.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
203.65
Special offer
Online exclusive
Ends: 28-Dec-2026
239.00
Save 35.35
Each
Chemical Identifiers
CAS166386-48-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White to yellow or pale cream
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only