2-(Methoxycarbonyl)benzeneboronic acid, 97%
2-(Methoxycarbonyl)benzeneboronic acid, 97%
2-(Methoxycarbonyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

2-(Methoxycarbonyl)benzeneboronic acid, 97%

CAS: 374538-03-1 | C8H9BO4 | 179.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17958.03
also known as L17958-03
Price (USD)
38.40
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
38.40
Each
Chemical Identifiers
CAS374538-03-1
IUPAC Name[2-(methoxycarbonyl)phenyl]boronic acid
Molecular FormulaC8H9BO4
InChI KeyODAXNYMENLFYMY-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)100.0-109.0?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only