4-Piperidinemethanol, 97%
4-Piperidinemethanol, 97%
4-Piperidinemethanol, 97%
4-Piperidinemethanol, 97%
Thermo Scientific Chemicals

4-Piperidinemethanol, 97%

CAS: 6457-49-4 | C6H13NO | 115.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L17964.14
also known as L17964-14
Price (USD)
263.65
Special offer
Online exclusive
Ends: 30-Jun-2026
310.00
Save 46.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
263.65
Special offer
Online exclusive
Ends: 30-Jun-2026
310.00
Save 46.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS6457-49-4
IUPAC Name4-(hydroxymethyl)piperidin-1-ium
Molecular FormulaC6H14NO
InChI KeyXBXHCBLBYQEYTI-UHFFFAOYSA-O
SMILESOCC1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Water Content (Karl Fischer Titration)≤1.5%
Melting Point (clear melt)54.0-63.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only